Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265876.1 | NP_001252805.1 | 401 | 99 | ||
GenBank mRNA | JU334545.1 | - | 406 | 99 | ||
JU475631.1 | - | 415 | 99 | |||
JV047082.1 | - | 401 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |