Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194267.2 | NP_001181196.1 | 337 | 150 | ||
XM_001104266.4 | XP_001104266.1 | 561 | 150 | |||
XM_015112908.2 | XP_014968394.1 | 413 | 150 | |||
GenBank mRNA | JU332441.1 | - | 478 | 150 | ||
JU472973.1 | - | 467 | 150 | |||
JV045208.1 | - | 439 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |