Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266725.1 | NP_001253654.1 | 388 | 68 | ||
XM_028846754.1 | XP_028702587.1 | 541 | 68 | |||
GenBank mRNA | JU323192.1 | - | 449 | 68 | ||
JU471942.1 | - | 437 | 68 | |||
JV044246.1 | - | 388 | 68 | |||
JV639010.1 | - | 494 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |