Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267785.1 | NP_001254714.1 | 373 | 127 | ||
XM_015147245.2 | XP_015002731.2 | 486 | 127 | |||
GenBank mRNA | JU337257.1 | - | 388 | 127 | ||
JU477964.1 | - | 373 | 127 | |||
JV727579.1 | - | 399 | 127 | |||
KT763477.1 | - | 366 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |