Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267539.1 | NP_001254468.1 | 202 | 94 | ||
GenBank mRNA | JU321066.1 | - | 285 | 94 | ||
JV045569.1 | - | 202 | 94 | |||
JV529667.1 | - | 272 | 94 | |||
JV602437.1 | - | 286 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |