Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193491.1 | NP_001180420.1 | 973 | 63 | ||
XM_015123082.2 | XP_014978568.1 | 1317 | 63 | |||
XM_015123083.2 | XP_014978569.1 | 2131 | 63 | |||
XM_015123084.1 | XP_014978570.1 | 1116 | 63 | |||
XM_015123085.2 | XP_014978571.1 | 1096 | 63 | |||
GenBank mRNA | JU327645.1 | - | 815 | 63 | ||
JU473916.1 | - | 1072 | 63 | |||
JV045867.1 | - | 1036 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |