Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079696.1 | NP_001073164.1 | 617 | 64 | ||
XM_015132841.2 | XP_014988327.1 | 794 | 64 | |||
XM_015132842.2 | XP_014988328.1 | 769 | 64 | |||
XM_015132843.2 | XP_014988329.1 | 725 | 64 | |||
GenBank mRNA | EF101562.1 | - | 617 | 64 | ||
JU475682.1 | - | 666 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |