Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194477.1 | NP_001181406.1 | 491 | 94 | ||
GenBank mRNA | JU470851.1 | - | 398 | 94 | ||
JV046077.1 | - | 531 | 94 | |||
JV641233.1 | - | 534 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |