Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205127.2 | NP_001192056.2 | 3227 | 84 | ||
XM_015112348.2 | XP_014967834.1 | 2786 | 84 | |||
XM_015112349.2 | XP_014967835.2 | 2088 | 84 | |||
XM_015112350.2 | XP_014967836.1 | 624 | 84 | |||
GenBank mRNA | CN643847.1 | - | 673 | 84 | ||
JU471546.1 | - | 442 | 84 | |||
JV045048.1 | - | 336 | 84 | |||
JV588048.1 | - | 3444 | 84 | |||
JV622917.1 | - | 3124 | 84 |