Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260729.1 | NP_001247658.1 | 701 | 106 | ||
XM_015142767.2 | XP_014998253.1 | 553 | 106 | |||
GenBank mRNA | JU332405.1 | - | 874 | 106 | ||
JU473866.1 | - | 693 | 106 | |||
JV045838.1 | - | 701 | 106 | |||
JV491365.1 | - | 753 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |