Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261493.1 | NP_001248422.1 | 319 | 149 | ||
XM_015133735.2 | XP_014989221.1 | 838 | 149 | |||
XM_015133736.2 | XP_014989222.1 | 694 | 149 | |||
GenBank mRNA | JU334836.1 | - | 271 | 149 | ||
JU472937.1 | - | 319 | 149 | |||
JV618615.1 | - | 320 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |