Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083949.1 | NP_001077418.1 | 182 | 79 | ||
XM_015152842.2 | XP_015008328.2 | 455 | 79 | |||
GenBank mRNA | EF467665.1 | - | 182 | 79 | ||
EF467666.1 | - | 182 | 79 | |||
JV638063.1 | - | 259 | 79 | |||
MH188460.1 | - | 182 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |