Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266887.2 | NP_001253816.1 | 441 | 81 | ||
GenBank mRNA | JU320160.1 | - | 568 | 81 | ||
JU473542.1 | - | 578 | 81 | |||
JV044890.1 | - | 442 | 81 | |||
JV639468.1 | - | 615 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |