Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194542.1 | NP_001181471.1 | 757 | 124 | ||
GenBank mRNA | JU322299.1 | - | 603 | 124 | ||
JU472469.1 | - | 525 | 124 | |||
JV045317.1 | - | 622 | 124 | |||
JV602177.1 | - | 721 | 124 | |||
JV636505.1 | - | 674 | 124 | |||
JV668253.1 | - | 744 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |