Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265840.2 | NP_001252769.1 | 209 | 59 | ||
GenBank mRNA | JU321435.1 | - | 203 | 59 | ||
JU472311.1 | - | 196 | 59 | |||
JV045676.1 | - | 209 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |