Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271653.1 | NP_001258582.1 | 738 | 121 | ||
NM_001271654.1 | NP_001258583.1 | 735 | 121 | |||
NM_001271655.1 | NP_001258584.1 | 675 | 121 | |||
NM_001271656.1 | NP_001258585.1 | 661 | 121 | |||
NM_001271657.1 | NP_001258586.1 | 658 | 121 | |||
XM_015113171.2 | XP_014968657.2 | 723 | 121 | |||
XM_015113175.2 | XP_014968661.2 | 727 | 121 | |||
GenBank mRNA | DQ423379.1 | - | 576 | 121 | ||
DQ423380.1 | - | 555 | 121 | |||
EF396932.1 | - | 675 | 121 | |||
EF396933.1 | - | 661 | 121 | |||
EF396934.1 | - | 658 | 121 | |||
EF396935.1 | - | 738 | 121 | |||
EF396936.1 | - | 735 | 121 | |||
EF396937.1 | - | 738 | 121 | |||
EF396938.1 | - | 661 | 121 | |||
EF396939.1 | - | 735 | 121 | |||
EF396940.1 | - | 738 | 121 | |||
HQ423386.1 | - | 576 | 121 | |||
HQ423387.1 | - | 576 | 121 | |||
HQ423388.1 | - | 576 | 121 | |||
JQ038004.2 | - | 684 | 121 | |||
JQ038005.1 | - | 684 | 121 | |||
JQ038006.1 | - | 684 | 121 | |||
JV709540.1 | - | 246 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta Inventoried |