Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117183.4 | XP_001117183.3 | 679 | 61 | ||
XM_015112746.2 | XP_014968232.2 | 699 | 61 | |||
XM_028827216.1 | XP_028683049.1 | 748 | 61 | |||
XM_028827222.1 | XP_028683055.1 | 397 | 61 | |||
XM_028827225.1 | XP_028683058.1 | 441 | 61 | |||
GenBank mRNA | JU325040.1 | - | 357 | 61 | ||
JU471931.1 | - | 608 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |