Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMU-DOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194682.3 | NP_001181611.2 | 185 | 69 | ||
GenBank mRNA | JU485415.1 | - | 150 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |