Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194836.2 | NP_001181765.1 | 595 | 80 | ||
XM_001118584.4 | XP_001118584.2 | 539 | 80 | |||
XM_028840603.1 | XP_028696436.1 | 590 | 80 | |||
GenBank mRNA | JU336211.1 | - | 565 | 80 | ||
JU477055.1 | - | 440 | 80 | |||
JV602161.1 | - | 567 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |