Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257551.1 | NP_001244480.1 | 486 | 125 | ||
XM_028826783.1 | XP_028682616.1 | 262 | 125 | |||
GenBank mRNA | JU335734.1 | - | 486 | 125 | ||
JU476623.1 | - | 450 | 125 | |||
JV047812.1 | - | 418 | 125 | |||
JV720273.1 | - | 498 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |