Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC711810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112727.2 | XP_014968213.1 | 502 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |