Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194568.1 | NP_001181497.1 | 379 | 157 | ||
XM_015112722.1 | XP_014968208.1 | 438 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |