Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261744.2 | NP_001248673.1 | 665 | 104 | ||
XM_015123385.2 | XP_014978871.1 | 1196 | 104 | |||
GenBank mRNA | JU321359.1 | - | 663 | 104 | ||
JV044634.1 | - | 666 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |