Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC697687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085444.4 | XP_001085444.1 | 411 | 156 | ||
GenBank mRNA | JV523673.1 | - | 275 | 156 | ||
JV699398.1 | - | 256 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta Inventoried |