Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085284.4 | XP_001085284.2 | 581 | 139 | ||
XM_001085512.4 | XP_001085512.1 | 289 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |