Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001118889.2 | NP_001112361.1 | 247 | 63 | ||
NM_001346420.1 | NP_001333349.1 | 375 | 63 | |||
GenBank mRNA | CN643832.1 | - | 148 | 63 | ||
JV046824.1 | - | 181 | 63 | |||
JV465885.1 | - | 178 | 63 | |||
JV602446.1 | - | 375 | 63 | |||
JV649198.1 | - | 246 | 63 | |||
JV667383.1 | - | 301 | 63 | |||
JV681449.1 | - | 233 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |