Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257806.1 | NP_001244735.1 | 175 | 85 | ||
GenBank mRNA | JU334871.1 | - | 175 | 85 | ||
JU472989.1 | - | 162 | 85 | |||
JV044650.1 | - | 177 | 85 | |||
JV653289.1 | - | 141 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |