Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266904.1 | NP_001253833.1 | 617 | 82 | ||
XM_015129798.2 | XP_014985284.1 | 956 | 82 | |||
GenBank mRNA | JU334168.1 | - | 617 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |