Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127877.2 | XP_014983363.1 | 296 | 121 | ||
GenBank mRNA | JU334824.1 | - | 265 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |