Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126450.2 | XP_014981936.1 | 548 | 101 | ||
XM_015126451.2 | XP_014981937.1 | 396 | 101 | |||
XM_028841542.1 | XP_028697375.1 | 321 | 101 | |||
GenBank mRNA | JV064396.1 | - | 139 | 101 | ||
JV472713.1 | - | 131 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |