Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115254.3 | XP_001115254.1 | 148 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |