Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103809.3 | XP_001103809.2 | 5063 | 65 | ||
XM_028840149.1 | XP_028695982.1 | 7090 | 65 | |||
GenBank mRNA | JU327298.1 | - | 4715 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |