Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261646.1 | NP_001248575.1 | 748 | 110 | ||
XM_028850205.1 | XP_028706038.1 | 1225 | 110 | |||
GenBank mRNA | JV044147.1 | - | 748 | 110 | ||
JV635637.1 | - | 753 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |