Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148315.2 | XP_015003801.1 | 146 | 85 | ||
GenBank mRNA | JU335916.1 | - | 81 | 85 | ||
JU472153.1 | - | 99 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |