Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093244.4 | XP_001093244.1 | 236 | 79 | ||
XM_028839637.1 | XP_028695470.1 | 241 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |