Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTHL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194051.3 | NP_001180980.2 | 635 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |