Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266795.2 | NP_001253724.1 | 272 | 130 | ||
GenBank mRNA | JU336704.1 | - | 259 | 130 | ||
JU477493.1 | - | 257 | 130 | |||
JV048438.1 | - | 276 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |