Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193727.2 | NP_001180656.2 | 1582 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |