Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143229.2 | XP_014998715.2 | 2049 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |