Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098246.3 | XP_001098246.1 | 134 | 114 | ||
GenBank mRNA | JV704691.1 | - | 106 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |