Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124842.2 | XP_014980328.2 | 1474 | 119 | ||
GenBank mRNA | JV589669.1 | - | 124 | 119 | ||
JV624689.1 | - | 294 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |