Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093499.4 | XP_001093499.2 | 435 | 141 | ||
XM_028826663.1 | XP_028682496.1 | 435 | 141 | |||
XM_028826664.1 | XP_028682497.1 | 433 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |