Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261485.1 | NP_001248414.1 | 713 | 63 | ||
GenBank mRNA | JU474533.1 | - | 713 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |