Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARBI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261506.2 | NP_001248435.1 | 843 | 118 | ||
XM_015114202.2 | XP_014969688.1 | 1190 | 118 | |||
XM_015114203.2 | XP_014969689.1 | 1195 | 118 | |||
XM_015114204.2 | XP_014969690.1 | 892 | 118 | |||
GenBank mRNA | JU321448.1 | - | 870 | 118 | ||
JU471418.1 | - | 843 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |