Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193430.1 | NP_001180359.1 | 122 | 108 | ||
XM_028838503.1 | XP_028694336.1 | 211 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |