Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOSPHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092665.4 | XP_001092665.1 | 292 | 109 | ||
XM_015119340.2 | XP_014974826.1 | 126 | 109 | |||
XM_028836389.1 | XP_028692222.1 | 126 | 109 | |||
XM_028836392.1 | XP_028692225.1 | 1787 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |