Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084230.4 | XP_001084230.2 | 973 | 130 | ||
XM_015126337.2 | XP_014981823.1 | 411 | 130 | |||
GenBank mRNA | JV672662.1 | - | 445 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |