Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC704334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133039.2 | XP_014988525.1 | 1544 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |