Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087365.4 | XP_001087365.1 | 1583 | 155 | ||
XM_015143725.2 | XP_014999211.1 | 1583 | 155 | |||
GenBank mRNA | JU484364.1 | - | 289 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |