Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261718.1 | NP_001248647.1 | 682 | 116 | ||
XM_015128385.2 | XP_014983871.1 | 761 | 116 | |||
XM_015128386.2 | XP_014983872.1 | 1050 | 116 | |||
XM_015128387.2 | XP_014983873.1 | 761 | 116 | |||
XM_028841760.1 | XP_028697593.1 | 659 | 116 | |||
GenBank mRNA | JU322282.1 | - | 733 | 116 | ||
JV047122.1 | - | 682 | 116 | |||
JV652072.1 | - | 761 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |